Ethyl acetate 99,5 ACS zA, General Laboratory Use
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
Synonyms:
Acetic acid ethyl ester
LP100-19
LP100-4
LP100-4P
CABJLP100D19EA
1107.49
CAD
CABJLP100D19
CABJLP100-4
BJLP100-204
BJLP100-4P
Ethyl acetate 99,5 ACS zA, General Laboratory Use
Ethyl acetate
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Formula:
CH₃COOC₂H₅ MW: 88.11 g/mol Boiling Pt: 77.1 Melting Pt: -83 Density: 0.9006 g/cm³ (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009171 CAS Number: 141-78-6 EINECS: 205-500-4 UN: 1173 ADR: 3 II REACH: 01-2119475103-46 Merck Index: 13,03792 |
|
Specification Test Results
Assay by GC Analysis | 99.5 % min. |
Color (APHA) | 10 max. |
Residue after Evaporation | 0.003 % max. |
Substances Darkened by Sulfuric Acid | Passes test |
Titrable Acid | 0.9 µeq/g max. |
Water by Karl Fischer Titration | 0.2 % max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...