Ethyl acetate ≥99.5% ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
Synonyms:
Acetic acid ethyl ester
AH100-4PC
AH100-4
CABJAH100-4PCCS
1126.62
CAD
CABJAH100-4PC
CABJAH100-4
Ethyl acetate ≥99.5% ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Ethyl acetate
Meets USP Reagent Specifications
- ACS certified
- Lot-to-lot consistency
- HPLC grade controlled for UV absorbance, GC assay, water content and non-volatile residue
Formula:
CH₃COOC₂H₅ Boiling Pt: 11.5 %(V) Melting Pt: –84 °C Density: 0.9006 g/cm³ (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009171 CAS Number: 141-78-6 EINECS: 205-500-4 UN: 1173 ADR: 3,II REACH: 01-2119475103-46 Merck Index: 13,03792 |
|
Specification Test Results
Color (APHA) | 10 max. |
GC Analysis | 99.5 % min. |
Peroxides (as H₂O₂) at time of packaging | 2 mg/L max. |
Refractive Index (20ºC) | 1.3720 - 1.3726 |
Residue | 3 mg/L max. |
Substances Darkened by Sulfuric Acid | Passes test |
Titrable Acid | 0.9 µeq/g max. |
UV Cutoff | 256 nm max. |
Water by Karl Fischer Titration | 0.03 % max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...