Diethyl ether ≥99.9% (excluding ethanol) stabilized, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
Stabilization:
Stabilised with ethanol 2%
106-1L
106-4
CABJ106-1LCS
812.37
CAD
CABJ106-1L
CABJ106-4
Diethyl ether ≥99.9% (excluding ethanol) stabilized, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Diethyl ether
For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.
- Minimal UV contaminants
- High GC purity
- Low water content
- Outstanding lot-to-lot consistency
Formula:
(CH₃CH₂)₂O Boiling Pt: 35 °C Melting Pt: –117.4 °C Density: 0.713 g/cm³ (20 °C) Flash Pt: –45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011646 CAS Number: 60-29-7 EINECS: 200-467-2 UN: 1155 ADR: 3,I REACH: 01-2119535785-29 Merck Index: 12,03852 |
|
Specification Test Results
Electron Capture GC | 10 ng/L max. |
Water by Karl Fischer Titration | 0.03 % max. |
GC Analysis (excluding ethanol) | 99.9 % min. |
Peroxides (as H₂O₂) at time of packaging | 1 ppm max. |
Refractive Index (20ºC) | 1.3523 - 1.3533 |
Residue | 1 mg/L max. |
UV Absorbance @ 250 nm | 0.08 AU max. |
UV Absorbance @ 275 nm | 0.01 AU max. |
UV Absorbance @ 300 nm | 0.005 AU max. |
UV Cutoff | 215 nm max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...