Ethyl acetate ≥99.9%, B&J Brand™ ACS for HPLC, for spectrophotometry, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Acetic acid ethyl ester

100-4 NP100-20 100-4
CABJ100-4CS 1210.68 CAD
CABJ100-4 CABJNP100-20 BJ100-4
Ethyl acetate ≥99.9%, B&J Brand™ ACS for HPLC, for spectrophotometry, Burdick & Jackson™
Ethyl acetate

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: CH₃COOC₂H₅
Boiling Pt: 77 °C
Melting Pt: –84 °C
Density: 0.9006 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Electron Capture GC 10 ng/L max.
Water by Karl Fischer Titration 0.03 % max.
GC Analysis 99.9 % min.
Refractive Index (20ºC) 1.3719 - 1.3727
Residue 1 mg/L max.
UV Absorbance @ 275 nm 0.05 AU max.
UV Absorbance @ 300 nm 0.03 AU max.
UV Absorbance @ 325 nm 0.005 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 256 nm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR