Nalgene® Disposable Erlenmeyer Flasks, PETG, Sterile, Plain Bottom, Thermo Scientific
Supplier: Thermo Fisher Scientific
4112-0250 4112-0500 4112-2000 4112-0125 4112-1000 4112-2800
73520-396CS 274.24 CAD
73520-396 73520-398 73520-400 73520-462 73520-464 82031-372
Nalgene® Disposable Erlenmeyer Flasks, PETG, Sterile, Plain Bottom, Thermo Scientific
Flasks Erlenmeyer Flasks
Graduated flasks are made of light, crystal-clear PETG plastic with easy-to-open polyethylene screw closures.
  • Disposing of flasks reduces chance of cross-contamination
  • Non-pyrogenic, non-cytotoxic
  • Sterilized to SAL 10-6
The leak-proof high-density polyethylene screw closures open to vent with 1/4 turn.
Ideal for shaker and suspension cell culture, media preparation, or storage
Packaging: Individually packaged for easy storage.