Nalgene® Erlenmeyer Flasks, Teflon® FEP, with Screw Cap, Thermo Scientific
Supplier: Thermo Fisher Scientific
4106-0125 4106-0250
29136-740CS 1094.42 CAD
29136-740 29136-762
Nalgene® Erlenmeyer Flasks, Teflon® FEP, with Screw Cap, Thermo Scientific
Flasks Erlenmeyer Flasks
Flasks are unbreakable, nearly transparent and non-contaminating.
  • With convenient Tefzel® EFTE cap
  • Can be boiled in nitric acid for trace metals work
Teflon resists virtually all chemicals and withstand temperatures from -270°C to +205°C. Can be autoclaved or chemically sterilized. Before autoclaving, set cap on top of flask without engaging threads.