Diethyl ether, anhydrous ≥99.0% stabilized, ARISTAR® ACS, VWR Chemicals BDH®

Supplier: BDH

Synonyms: Ether2,2'-OxybisethaneEthoxyethaneEthyl Ether2,2'-Oxydiethane

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

203500-23503 203500-80405 203500-80405
CABDH3116-1L-1CS 1683.07 CAD
BDH3116-55GL CABDH3116-1L-1 BDH3116-1L-1
Diethyl ether, anhydrous ≥99.0% stabilized, ARISTAR® ACS, VWR Chemicals BDH®
Diethyl ether
Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.7135 g/cm³ (20 °C)
Flash Pt: –40 °C
MDL Number: MFCD00011646
CAS Number: 60-29-7
UN: 1155
ADR: 3,I
Merck Index: 12,03852
https://ca.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg

Order Now

Specification Test Results

Peroxide (as H₂O₂) 1 ppm max.
Preservative (BHT) Level Record only
Residue After Evaporation 0.001% max.
Titrable Acid 0.0002 meq/g max.
Water (H₂O) 0.03% max.
Assay ((CH₃CH₂)₂O) 99.0% min.
Carbonyl (as HCHO) 0.001% max.
Color (APHA) 10 max.
Alcohol (CH₃CH₂OH) To pass test (0.05%)
Identification:IR To Pass Test
Manufacture Date Record Actual
Use by Date Record Actual

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR