ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
Synonyms:
Reagent alcohol
BDH1164-19L
BDH1164-4LP
BDH1164-19LEA
656.16
CAD
BDH1164-19L
BDH1164-4LP
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Alcohol
Meets ACS specifications for general use.
Storage Temperature:
Ambient |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 EINECS: 200-578-6 UN: 1170 ADR: 3 III |
Specification Test Results
Appearance | Clear, colorless liquid |
Water | 29.0-31.0% |
Residue | Less than 0.001% (0.0010 g/115mL) |
GC Purity Ethanol | 63.0-64.0% |
GC Purity Methanol | 2.9-3.5% |
GC Purity Isopropyl Alcohol | 3.2-3.9% |
Color (APHA) | Less than 10 |
Specific gravity @ 20ºC | 0.879-0.895 |
Acidity | Less than 0.0025% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...