Ethanol denatured ACS (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Ethyl alcohol denatured

BDH1158-4LP BDH1158-19L
BDH1158-4LPCS 1335.96 CAD
BDH1158-4LP BDH1158-19L
Ethanol denatured ACS (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Ethanol denatured

  • Total alcohol: 95%
  • Ethyl alcohol: 85-87%
  • Denaturant: 4-5% methanol and 4.5–5.5% 2-propanol.


Ordering information: The purchase of Reagent Alcohol in containers larger than 4 liters requires a Letterhead Permit approval by the United States Treasury Tax and Trade Bureau (TTB).

Formula: H₃CCH₂OH
MW: 46.07 g/mol
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Water 4.0–6.0%
Residue Less than 0.001% (0.0010g/122mL)
GC Purity Ethanol 85.0–87.0%
GC Purity Methanol 4.5–5.0%
GC Purity Isopropyl Alcohol 4.5–5.5%
Color (APHA) Less than 10
Specific gravity @ 20ºC 0.806-0.813
Acidity Less than 0.0025%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR