Reagent alcohol ACS (MeOH+ IPA), VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
Synonyms:
Reagent alcohol
BDH1156-1LP
BDH1156-4LP
BDH1156-5GL
BDH1156-19L
BDH1156-1LPCS
414.14
CAD
BDH1156-1LP
BDH1156-4LP
BDH1156-5GL
BDH1156-19L
Reagent alcohol ACS (MeOH+ IPA), VWR Chemicals BDH®
Alcohol
Meets ACS specifications for general use.
- Total alcohol: 99.5%
- Ethyl alcohol: 89-91%
- Denaturant: 4-6% methyl alcohol and 4-6% 2-propanol
Ordering information: The purchase of Reagent Alcohol in containers larger than 4 liters requires a Letterhead Permit approval by the United States Treasury Tax and Trade Bureau (TTB).
Formula:
H₃CCH₂OH Storage Temperature: Ambient |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 EINECS: 200-578-6 UN: 1170 ADR: 3 II |
Specification Test Results
Appearance | Clear, colorless liquid |
Water (K. F.) | Less than 0.50% |
Residue | Less than 0.001% (0.0010 g/124 mL) |
Assay (Ethanol + Methanol) | 94.0 – 96.0 % |
Assay (Isopropyl Alcohol) | 4.0-6.0% |
Color (APHA) | Less than 10 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...