Ethylene glycol ≥99% ACS, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
|
Warning
|
Synonyms:
Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol
BDH1125-1LP
BDH1125-4LG
BDH1125-4LP
BDH1125-1LPCS
478.62
CAD
BDH1125-1LP
BDH1125-4LG
BDH1125-4LP
Ethylene glycol ≥99% ACS, Laboratory Reagent, VWR Chemicals BDH®
Ethylene glycol
Clear, colorless liquid.
Formula:
HOCH₂CH₂OH MW: 62.07 g/mol Boiling Pt: 196…198 °C (1013 hPa) Melting Pt: -13 Density: 1.115 g/cm³ (20 °C) Flash Pt: 116 Storage Temperature: Refrigerator |
MDL Number:
MFCD00002885 CAS Number: 107-21-1 EINECS: 203-473-3 REACH: 01-2119456816-28 Merck Index: 13,03832 |
|
Specification Test Results
Appearance | Clear, colorless liquid |
Water (K.F.) | Less than 0.20% |
GC Purity | Greater than 99.0% |
Color (APHA) | Less than 10 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...