Corning® Polycarbonate Erlenmeyer Flasks with Screw Caps
Supplier: Corning
431252 431256 431253 431255 430421 430183 430422 431144 431145 431143 431146 431147 431404 431405 431402 431403 431406 431407 431408 431401 431684 431685
17777-100CS 568.74 CAD
17777-100 17777-102 17777-104 17777-106 29152-146 29152-168 29152-180 29445-056 29445-058 29445-060 29445-062 29445-064 89089-716 89089-718 89089-750 89089-752 89089-758 89089-760 89089-810 89089-812 10098-584 10098-586
Corning® Polycarbonate Erlenmeyer Flasks with Screw Caps
Flasks Erlenmeyer Flasks
PC, optically clear.
  • Baffled or plain bottom options in all sizes (125 ml to 3 L)
  • Moulded-in graduations for accuracy
  • Vent cap option for continuous gas exchange while ensuring sterility and preventing leakage
  • Individually packaged and radiation sterilised for ease of use
  • Certified non-pyrogenic
Corning® baffled and plain Erlenmeyer flasks are ideal for shaker culture applications and storage.
Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.