Sartobind® Lab IEX Membrane Adsorbers
Supplier: Sartorius
93IEXD42DB12V 93IEXQ42BC12 93IEXQ42DB12V 93IEXQ42GB12A 93IEXS42BC12 93IEXS42DB12V 93IEXS42GB12A
CA76795-406PK 289.15 CAD
CA76795-406 CA76795-408 CA76795-410 CA76795-412 CA76795-414 CA76795-416 CA76795-418
Sartobind® Lab IEX Membrane Adsorbers
Filters Syringe Filters
Small-scale laboratory units for high speed ion exchange purifications, which can be re-used hundreds of times.
  • High capacity anion or cation exchange ligands
  • Rapid, equipment-free purification by syringe
  • Adapters included for use with FPLC systems
  • Maximum Operating Pressure: 0.6 MPa
Process-Ready Sartobind® Membrane Adsorbers Sartobind® Lab membrane adsorbers for ion exchange chromatography are ready-to-use units in a syringe filter format. They are based on the macroporous Sartobind® membranes, with anion or cation exchange ligands covalently bound to the entire internal surface of the pores. This results in a media chromatography matrix with superior flow rates and high binding capacity. Covalent linkage of ion exchange ligands to the membrane is very stable, allowing for many cycles of re-use without loss of binding capacity.
In addition, these units utilize the same membrane adsorber platform as found in Sartobind® capsules and cassettes, making them ideal for research and development work, prior to convenient scale-up for manufacturing. Ultimate VersatilitySartobind® Lab units for ion exchange are available with strong or weak anion (Q and D) and strong cation (S) exchangers, and in three bed volumes, ranging from 0.41 to 2.8 ml, corresponding to dynamic binding capacities between 12 and 80 mg protein per unit. Equipped with standard Luer lock connectors, Sartobind® Lab units enable simple, equipment-free handling by syringe, or can be easily connected to FPLC systems for automated purifications. The units are compatible with most common chromatography buffers and can be re-used 1000 times, ensuring high-cost efficiency.