Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
BDH1121-19L
BDH1121-1LPC
BDH1121-4LPC
BDH1121-19LEA
1175.33
CAD
BDH1121-19L
BDH1121-1LPC
BDH1121-4LPC
Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Diethyl ether
Suitable for organic synthesis, cleaning, and prep scale synthesis. Contains ~1ppm BHT as preservative.
Formula:
(CH₃CH₂)₂O Boiling Pt: 35 °C (1013 hPa) Melting Pt: –123 °C Density: 0.7135 g/cm³ (20 °C) Flash Pt: –40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011646 CAS Number: 60-29-7 EINECS: 200-467-2 UN: 1155 ADR: 3,I REACH: 01-2119535785-29 Merck Index: 12,03852 |
|
Specification Test Results
Appearance | Clear, colorless liquid |
Water (K.F.) | Less than 0.030% |
Residue | Less than 0.001% (0.0010 g/141 ml) |
GC Purity | Greater than 99.0% |
Peroxides (TiCl₄; mg/L) | Less than 1.0 mg/L as H₂O₂ (Typical - Not detected (less than 0.1 mg/L)) |
Peroxides (TiCl₄; mg/Kg) | Less than 1.0 mg/Kg as H₂O₂ (Typical - Not detected (less than 0.1 mg/Kg)) |
Color (APHA) | Less than 10 |
Titrable acid | Less than 0.2 meq/g (Not more than 0.30 ml of 0.01N sodium hydroxide) |
Carbonyl (as HCHO) | Less than 0.001% (The total diffusion current should not be greater than ½ that of sample plus aldehyde) |
Alcohol (as CH₂CH₃OH) | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...