Ethanol denatured SDA-1 formula, Epredia™ HiPur™ for histology (denatured with 4% methanol)

Supplier: RICHARD-ALLAN SCIENTIFIC CA

9200-5 9200-1
CA48218-736EA 628.78 CAD
CA48218-736 CA48218-740
Ethanol denatured SDA-1 formula, Epredia™ HiPur™ for histology (denatured with 4% methanol)
Ethanol denatured

Epredia™ HiPur™ Dedyrants are histological grade and anhydrous. Denatured with ethyl acetate, methanol, methyl isobutyl ketone, and heptane.


  • Denaturant: 4% methanol
  • Maximum of 0.1% water
  • Maximum residue content of 0.003%


Meets spectrophotometric grade specifications.

Formula: H₃CCH₂OH
MDL Number: MFCD00003568
CAS Number: 64-17-5

Order Now

Specification Test Results

Assay 90–92%
Residue 0.003% max.
Water 0.1% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR