3,4-Dimethylhippuric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3,4-Dimethylhippuric acid, 2-(3,4-Dimethylbenzamido)acetic acid, N-(3,4-Dimethylbenzoyl)glycine

D2241-5G D2241-1G
TCD2241-5GEA 6880.9 CAD
TCD2241-5G TCD2241-001G
3,4-Dimethylhippuric acid ≥98.0% (by titrimetric analysis)
3,4-Dimethylhippuric acid

CAS Number: 23082-12-4
MDL Number: MFCD00191414
Molecular Formula: C11H13NO3
Molecular Weight: 207.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 160


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-(3,4-Dimethylbenzoyl)glycine

Formula: C₁₁H₁₃NO₃
MW: 207.23 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00191414
CAS Number: 23082-12-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e86e3457f0bf91ec7707bb34baf8924259a967e&vid=0e86e3457f0bf91ec7707bb34baf8924259a967e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR