KIMAX® Erlenmeyer Flasks with [ST] Joint, Graduated, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
26510-2000 26510-50 26510-125 26510-250 26510-500 26510-1000
89001-340CS 206.01 CAD
89001-340 89000-788 89001-332 89001-334 89001-336 89001-338
KIMAX® Erlenmeyer Flasks with [ST] Joint, Graduated, Kimble Chase
Flasks Erlenmeyer Flasks
Flasks are graduated with white scales to indicate approximate volumes and feature a full length [ST] ground glass neck finish.
  • Tops are reinforced and tooled with a rounded finish, containing more glass to give them maximum mechanical strength
  • Body is thick-walled, with a long tapered outside contour to minimize chipping when struck or rubbed together
  • With durable white ceramic enamel capacity scale
Designed from ASTM Specification E1404, Type II, Class 1 requirements.
Manufactured from 33 expansion, low extractable borosilicate glass conformingMore Product Information