2,3-Dihydrobenzo[b]furan-7-carboxylic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,3-Dihydrobenzofuran-7-carboxylic acid

D2446-5G D2446-1G
TCD2446-5GEA 505.76 CAD
TCD2446-5G TCD2446-001G
2,3-Dihydrobenzo[b]furan-7-carboxylic acid ≥98.0% (by titrimetric analysis)
2,3-Dihydrobenzo[b]furan-7-carboxylic acid

CAS Number: 35700-40-4
MDL Number: MFCD00671526
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 172


Other Product Names / Synonyms: 7-Coumarancarboxylic Acid

Formula: C₉H₈O₃
MW: 164.1580 g/mol
Melting Pt: 172 °C
MDL Number: MFCD00671526
CAS Number: 35700-40-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f83570da6ec36ddb99b3a9de0132a5a9b5618f35&vid=f83570da6ec36ddb99b3a9de0132a5a9b5618f35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR