2,6-Di-tert-butyl-4-(dimethylaminomethyl)phenol ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N,N-dimethyl-3,5-di-tert-butyl-4-hydroxybenzylamine

D2301-500G D2301-25G
TCD2301-500GEA 360.24 CAD
TCD2301-500G TCD2301-025G
2,6-Di-tert-butyl-4-(dimethylaminomethyl)phenol ≥98.0% (by HPLC, titration analysis)
2,6-Di-tert-butyl-4-(dimethylaminomethyl)phenol

CAS Number: 88-27-7
MDL Number: MFCD00026283
Molecular Formula: C17H29NO
Molecular Weight: 263.43
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 179
Melting point (°C): 94
Flash Point (°C): 138

Formula: C₁₇H₂₉NO
MW: 263.4183 g/mol
Boiling Pt: 179 °C
Melting Pt: 94 °C
Flash Pt: 138 °C
MDL Number: MFCD00026283
CAS Number: 88-27-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0dbf907974c5e9e410e38283e9782f16ebf13811&vid=0dbf907974c5e9e410e38283e9782f16ebf13811&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR