N,O-Dimethylhydroxylamine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D1899-500G D1899-25G D1899-100G
TCD1899-500GEA 589.34 CAD
TCD1899-500G TCD1899-025G TCD1899-100G
N,O-Dimethylhydroxylamine hydrochloride ≥98.0% (by titrimetric analysis)
N,O-Dimethylhydroxylamine hydrochloride

CAS Number: 6638-79-5
MDL Number: MFCD00012485
Molecular Formula: C2H7NO
Molecular Weight: 97.54
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 115


Product Categories: Synthetic Organic Chemistry, Amination
Other Product Names / Synonyms: N-Methoxy-N-methylamine Hydrochloride

Formula: C₂H₈ClNO
Melting Pt: 115 °C
MDL Number: MFCD00012485
CAS Number: 6638-79-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=141e476ad5f9f075c7121548a8e8e1ac57570c4d&vid=141e476ad5f9f075c7121548a8e8e1ac57570c4d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR