EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) ≥98.0%

Supplier: TCI America
Warning

Synonyms: N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochloride, 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDC hydrochloride, EDC, EDAC HCl, EDAC hydrochloride, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, WSC HCl, EDC HCl

D1601-250G D1601-5G D1601-25G
TCD1601-250GEA 1621.74 CAD
TCD1601-250G TCD1601-005G TCD1601-025G
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) ≥98.0%
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride

[Coupling Agent for Peptides Synthesis]
CAS Number: 25952-53-8
MDL Number: MFCD00012503
Molecular Formula: C8H17N3
Molecular Weight: 191.70
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 115
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis, Bioscience, Reagents for Oligosaccharide Synthesis, Crosslinkers
Other Product Names / Synonyms: 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide Hydrochloride, EDC.HCl, EDAC.HCl, EDCI.HCl

Formula: C₈H₁₇N₃·HCl
MW: 191.7 g/mol
Melting Pt: 115 °C
Storage Temperature: Freezer
MDL Number: MFCD00012503
CAS Number: 25952-53-8
EINECS: 247-361-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77f78571199ca07d4228bac14d24f73aadca8b9b&vid=2fe8f9e4cee273e5d81be15a76989c2e5da22d02&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR