1,3-Dimethyl-2-imidazolidinone ≥99.0%
Supplier: TCI America
|
|
Synonyms:
1,3-Dimethylimidazolidin-2-one
D1477-500ML
D1477-25ML
TCD1477-500MLEA
230
CAD
TCD1477-500ML
TCD1477-25ML
1,3-Dimethyl-2-imidazolidinone ≥99.0%
1,3-Dimethyl-2-imidazolidinone
CAS Number: 80-73-9
MDL Number: MFCD00003188
Molecular Formula: C5H10N2O
Molecular Weight: 114.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Melting point (°C): 8
Flash Point (°C): 120
Specific Gravity (20/20): 1.06
Other Product Names / Synonyms: N,N'-Dimethylethyleneurea, DMI, DMEU
Formula:
C₅H₁₀N₂O MW: 114.15 g/mol Boiling Pt: 108 °C Melting Pt: 8 Density: 1.06 (20 °C) Flash Pt: 120 °C |
MDL Number:
MFCD00003188 CAS Number: 80-73-9 EINECS: 201-304-8 UN: 2810 ADR: 6.1 III Merck Index: 13,03275 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...