(-)-Dibenzoyl-L-tartaric acid monohydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (2R,3R)-(-)-Di-O-benzoyl-tartaric acid monohydrate

D1354-25G D1354-500G
TCD1354-025GEA 127.98 CAD
TCD1354-025G TCD1354-500G
(-)-Dibenzoyl-L-tartaric acid monohydrate ≥98.0% (by titrimetric analysis)
(-)-Dibenzoyl-L-tartaric acid monohydrate

CAS Number: 62708-56-9
MDL Number: MFCD00149119
Molecular Formula: C18H14O8
Molecular Weight: 358.30
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 90
Specific rotation [a]20/D: -108.5 deg (C=1, EtOH)

Formula: C₁₈H₁₄O₈·1H₂O
MW: 376.33 (358.31 anhy)
Melting Pt: 90 °C
MDL Number: MFCD00149119
CAS Number: 62708-56-9
EINECS: 220-374-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5db1906a8468fd3531586ccbbd4c88f1c6ce6a27&vid=5db1906a8468fd3531586ccbbd4c88f1c6ce6a27&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR