1,12-Dibromododecane ≥98.0%
Supplier: TCI America
D1346-25G
D1346-500G
D1346-100G
TCD1346-025GEA
112.18
CAD
TCD1346-025G
TCD1346-500G
TCD1346-100G
1,12-Dibromododecane ≥98.0%
1,12-Dibromododecane
CAS Number: 3344-70-5
MDL Number: MFCD00000226
Molecular Formula: C12H24Br2
Molecular Weight: 328.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 215
Melting point (°C): 40
Flash Point (°C): 113
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Dodecamethylene Dibromide
Formula:
BrH₂C(CH₂)₁₀CH₂Br Boiling Pt: 215 °C Melting Pt: 40 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000226 CAS Number: 3344-70-5 EINECS: 222-096-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...