1,8-Dibromooctane ≥97.0%
Supplier: TCI America
D1064-25G
D1064-500G
D1064-100G
TCD1064-025GEA
110.6
CAD
TCD1064-025G
TCD1064-500G
TCD1064-100G
1,8-Dibromooctane ≥97.0%
1,8-Dibromooctane
CAS Number: 4549-32-0
MDL Number: MFCD00000277
Molecular Formula: C8H16Br2
Molecular Weight: 272.02
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 272
Flash Point (°C): 110
Freezing point (°C): 12
Specific Gravity (20/20): 1.47
Other Product Names / Synonyms: Octamethylene Bromide, Octamethylene Dibromide
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...