Nitri-Solve® Unsupported Nitrile Gloves, Showa
Supplier: Showa
727-07 727-08 727-09 727-10 727-11 737XL-10 717-07 717-08 717-09 717-10 717-11 730-09 730-11 717-11
32892-032CS 1163.09 CAD
32892-032 32892-033 32892-034 32892-035 32892-036 CA-32892-038 CA-32892-039 CA32892-027 CA32892-028 CA32892-029 CA32892-030 CA32892-031 CA32892-040 CA32892-041 CA32892-043 CA32892-045 CA32892-047 32892-031
Nitri-Solve® Unsupported Nitrile Gloves, Showa
Gloves
Hand and product protection for food processing, manufacturing, electronics, machine work, pharmaceutical, and laboratory applications.
  • Nitri-Solve® gloves are powder-free and have a case-hardened finish to enhance chemical and abrasion resistance
Easy donning and removal. Nonslip wet grip for safety. Hand-specific design with straight cuff for comfort. Choose from flock-lined and unlined styles in various lengths and thicknesses. Color: aqua-green.
Resist a broad range of chemicals where protection from intermittent contact or splash is needed. Resist abrasion, cuts, punctures, and snags.
Certifications: Gloves are made using only listed materials and levels outlined in 21 CFR 170-199.