AlphaTec® Solvex® Nitrile gloves, Ansell
Supplier: Ansell Canada
3717511070 3717511080 3717511090 3717511100 3717511110 3715511070 3715511080 3715511100 3715511110 3714511070 3714511080 3714511090 3714511100 3714511110 3716511070 3716511100 3716511110 3718511090 3718511100 3715511080 3715511090 3715511100 3715511110
CA-32890-128CS 555.5 CAD
CA-32890-128 CA-32890-130 CA-32890-132 CA-32890-134 CA-32890-136 CA32490-008 CA32490-009 CA32490-010 CA32490-011 CA32490-015 CA32890-088 CA32890-090 CA32890-092 CA32890-094 CA32890-096 CA32890-116 CA32890-118 CA32890-120 CA32890-122 CA32890-124 CA32890-140 CA32890-142 CA10798-910 CA10798-912 CA10798-932 CA10798-934
AlphaTec® Solvex® Nitrile gloves, Ansell
Gloves
Chemical-resistant gloves are suitable for chemical processing, food processing, aerospace and automotive degreasing, general manufacturing, electronics, and other applications.
  • Curved finger design and contoured palm to reduce fatigue
  • Embossed texture improves grip for easier handling of wet objects
  • Hand-specific
  • Powder-free
  • Straight cuffs
Tough nitrile compound provides long wear and good protection against abrasion, cuts, snags, and punctures. Strong resistance to degradation ensures that gloves will not weaken or swell. Optional cotton flock lining absorbs perspiration and makes donning and doffing easier. Gloves are free of protein allergens and will not promote contact dermatitis.
Protects against a wide range of chemicals, including aromatic solvents, cutting oils and coolants, petroleum solvents and animal fats.
Certifications: Component materials comply with FDA food handling requirements (21 CFR 177.2600).