Dodecylbenzene (mixture of branched chain isomers), hard type
Supplier: TCI America
D0988-25ML
D0988-500ML
TCD0988-025MLEA
45.82
CAD
TCD0988-025ML
TCD0988-500ML
Dodecylbenzene (mixture of branched chain isomers), hard type
Dodecylbenzene (mixture of branched chain isomers)
CAS Number: 25265-78-5
MDL Number: MFCD00008974
Molecular Formula: C18H30
Molecular Weight: 246.44
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 124
Specific Gravity (20/20): 0.87
Form: Clear Liquid
Color: Colorless
Formula:
C₁₈H₃₀ Density: 0.87 (20 °C) Flash Pt: 124 °C |
MDL Number:
MFCD00008974 CAS Number: 25265-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...