n-Dodecane ≥99.0%

Supplier: TCI America
Danger

D0968-25ML D0968-500ML
TCD0968-025MLEA 50.67 CAD
TCD0968-025ML TCD0968-500ML
n-Dodecane ≥99.0%
n-Dodecane

CAS Number: 112-40-3
MDL Number: MFCD00008969
Molecular Formula: C12H26
Molecular Weight: 170.34
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 216
Melting point (°C): -10
Flash Point (°C): 82
Specific Gravity (20/20): 0.75

Formula: H₃C(CH₂)₁₀CH₃
Boiling Pt: 216 °C
Melting Pt: –10 °C
Density: 0.75 (20 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00008969
CAS Number: 112-40-3
EINECS: 203-967-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e254196243654888d269bc1b6c259b10528ba5ea&vid=99af814b1093e32775d674af8cc229acf20eef0e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR