3,3'-Dimethylnaphthidine ≥97% (by titrimetric analysis) for chloride determination
Supplier: TCI America
|
|
Synonyms:
4,4'-Diamino-3,3'dimethyl-1,1'-binaphthyl, 3,3'-Dimethyl-[1,1'-binaphthalene]-4,4'-diamine, 4-(4-amino-3-methylnaphthalen-1-yl)-2-methylnaphthalen-1-amine
D0754-5G
D0754-1G
TCD0754-5GEA
1304.13
CAD
TCD0754-5G
TCD0754-001G
3,3'-Dimethylnaphthidine ≥97% (by titrimetric analysis) for chloride determination
3,3'-Dimethylnaphthidine
[for Colorimetric Determination of Cl in Water]
Form: Crystal
Melting point (°C): 232
Formula:
C₂₂H₂₀N₂ MW: 312.42 g/mol Melting Pt: 232 °C |
MDL Number:
MFCD00042706 CAS Number: 13138-48-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...