PYREX® Erlenmeyer Flasks, Graduated, Screw Cap, Corning
Supplier: Corning
4985-125 4985-250 4985-1L 4985-500 4985-50
89090-648CS 1534.43 CAD
89090-648 89090-786 89090-870 89090-874 89090-922
PYREX® Erlenmeyer Flasks, Graduated, Screw Cap, Corning
Flasks Erlenmeyer Flasks
Made of PYREX® borosilicate glass, with phenolic screw caps.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Graduated to show approximate capacity
  • With marking spot
Erlenmeyer flasks with phenolic screw caps are ideal for use as a shaker flask or for mixing, media prep and storage needs. Impact resistant caps are equipped with cemented-in, inert white rubber liners which are resistant to steam sterilization. Their uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance.