All-In-One Kitchenettes

Supplier: FELIX STORCH, INC./SUMMIT
C30EL C30ELBK C30ELGLASS C39ELGLASSBK CK36ELGLASS CK54SINKL CK54SINKR
77442-178EA 4131.05 CAD
77442-178 77442-180 77442-182 77442-184 77442-186 77442-188 77442-190
All-In-One Kitchenettes
Furniture Casework
All-in-one combination kitchenettes brings true convenience to smaller spaces with 2-burner cooktop, sink, storage cabinet, and refrigerator with a freezer compartment.

  • Refrigerator-freezer included
  • Sink and faucet
  • 115 V operation with pre-installed cords
  • Push-to-turn knobs
  • Indicator lights
  • Reversible doors
  • 100% CFC free

Refrigerant: R600a

Certifications: ENERGY STAR certified.

Packaging: Packaged and shipped in one box. Perfect for studio apartments, hotel rooms, dormitories, assisted living facilities, temporary kitchen facilities, and any other location in need of a low maintenance, fully featured compact kitchenette.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR