Dichloromethane ≥99.9% (by GC), ChromAR® for HPLC, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Methylene chloride

Management of Change (MOC) category = N

4879-06 MACR4879-10
MK487906CS 879.7 CAD
MK487906 CAMK487910
Dichloromethane ≥99.9% (by GC), ChromAR® for HPLC, for UV spectrophotometry, Macron Fine Chemicals™
Dichloromethane

Suitable for liquid chromatography and UV-spectrophotometry.

Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₂Cl₂) (by GC, corrected for water) ≥99.9 %
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.0003 %
ACS - Titrable Acid (meq/g) ≤0.0003
ACS - Free Halogens Passes Test
ACS - Water (H₂O)(by coulometry) ≤0.01 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 235 nm ≤1.0
Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm ≤0.12
Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm ≤0.04
Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 - 400 nm ≤0.01
Liquid Chromatography Suitability Absorbance Passes Test
Appearance (clear, colorless liquid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR