Dimethyl sulfoxide ≥99.9%, ChromAR® for HPLC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Methyl sulfoxideDMSODimethyl(oxido)sulfurMethyl sulphoxideMethanesulphinylmethaneDimethyl(oxido)sulphurMethanesulfinylmethane

Management of Change (MOC) category = N

2969-08 MACR2969-08
CAMK296908CS 5876.49 CAD
CAMK296908 MK296908
Dimethyl sulfoxide ≥99.9%, ChromAR® for HPLC, Macron Fine Chemicals™
Dimethyl sulphoxide

Suitable for Liquid Chromatography and UV-Spectrophotometry.


  • Filtered through a 0.2µm element


DMSO is an industrial chemical and is not to be sold or used as a drug for humans or animals.

Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18.45 °C
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 89 °C (closed cup)
MDL Number: MFCD00002089
CAS Number: 67-68-5
Merck Index: 13,03285
https://ca.vwr.com/stibo/web/std.lang.all/77/31/2297731.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay ((CH₃)₂SO) (by GC, corrected for water) ≥99.9 %
ACS - Residue after Evaporation ≤5 ppm
ACS - Titrable Acid (meq/g) ≤0.001
ACS - Water (H₂O)(by KF, coulometric) ≤0.05 %
Optical Absorbance (1-cm path vs water) - 270 nm ≤0.40
Optical Absorbance (1-cm path vs water) - 290 nm ≤0.18
Optical Absorbance (1-cm path vs water) - 310 nm ≤0.06
Optical Absorbance (1-cm path vs water) - 330 nm ≤0.02
Optical Absorbance (1-cm path vs water) - 350 - 400 nm ≤0.01
Appearance (clear, colorless liquid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR