BioX Vacuum Filtration Systems, Sterile
Supplier: TWD Scientific
BX15-4000 BX15-4150 BX15-4250 BX15-4500
77394-962CS 405.35 CAD
77394-962 77394-964 77394-966 77394-968
BioX Vacuum Filtration Systems, Sterile
Filters Bottle Top Filters
Systems for vacuum filtration of solutions including buffers, cell culture media, or other fluids.
  • PES membrane
  • 0.22 µm pore size
  • Lightweight heavy wall construction
  • Engraved graduations
  • Non-pyrogenic
Vacuum filters are very useful for the separation and purification of large volumes of samples.