N,N-Dimethylacetamide ≥99.5% (by GC), BAKER ANALYZED®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC

Management of Change (MOC) category = N

BAKRJ372-07 BAKRJ372-09
CAJTJ372-7EA 654.53 CAD
CAJTJ372-7 CAJTJ372-9
N,N-Dimethylacetamide ≥99.5% (by GC), BAKER ANALYZED®
N,N-Dimethylacetamide
Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 166.1
Melting Pt: -20
Density: 0.937 g/cm³ (20 °C)
Flash Pt: 70
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
UN: 2810
ADR: 6.1 III
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃CON(CH₃)₂) (by GC) ≥99.5 %
Density (g/mL) at 25°C 0.931 - 0.936
Residue after Ignition ≤0.005 %
pH of 20% Solution at 25°C 4.0 - 7.0
Water (H₂O) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR