Dextran, M.W. ~ 70,000, low fraction, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

BAKRG200-05 G200-05
CAJTG200-5EA 942.22 CAD
CAJTG200-5 JTG200-5
Dextran, M.W. ~ 70,000, low fraction, J.T.Baker®
Dextran

Average molecular weight is 60,000 to 90,000.

Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS Number: 9004-54-0

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR