Dithiothreitol (DTT, Cleland's reagent) ≥99.5%, Ultrapure Bioreagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

BAKRF780-03 BAKRF780-05
CAJTF780-1EA 723.15 CAD
CAJTF780-1 CAJTF780-2 CAJTF780-3 CAJTF780-5
Dithiothreitol (DTT, Cleland's reagent) ≥99.5%, Ultrapure Bioreagent, J.T.Baker®
Dithiothreitol (DTT, Cleland's reagent)
Formula: C₄H₁₀O₂S₂
Boiling Pt: ∼125 °C (1013 hPa)
Melting Pt: 42…44 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Keep refrigerated between 2-8°C (36-46°F) and store under nitrogen.
Assay (HSCH₂(CHOH)₂CH₂SH) ≥99.5 %
Absorbance of a 0.1M Solution (1-cm path vs water) at - 280 nm ≤0.060
Absorbance of a 0.1M Solution (1-cm path vs water) at - 260 nm ≤0.400
Appearance Passes Test
Melting Point 40 - 43 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR