Dichloromethane 99,5 GC, corrected for water content, PHOTREX® Spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Methylene chloride

M.O.C. Category N

BAKR9329-01 BAKR9329-03 9329-03
CAJT9329-1CS 3119.66 CAD
CAJT9329-1 CAJT9329-3 JT9329-3
Dichloromethane 99,5 GC, corrected for water content, PHOTREX® Spectrophotometry, J.T.Baker®
Dichloromethane

For spectrophotometry. UV cutoff 235nm. Lot analysis on label.

Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8
Melting Pt: -95
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₂Cl₂) (by GC, corrected for water) ≥99.5 %
Appearance Passes Test
ACS - Color (APHA) ≤10
ACS - Free Halogens Passes Test
ACS - Residue after Evaporation ≤0.001 %
ACS - Titrable Acid (µeq/g) ≤0.2
Ultraviolet Absorbance (1.00-cm cell vs. water) - 340-400 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm ≤0.04
Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm ≤0.35
Ultraviolet Absorbance (1.00-cm cell vs. water) - 235 nm ≤1.00
Water (by KF, coulometric) ≤0.02 %
Product Information (not specifications):
Windows of Infrared Transmittance (0.1-mm path, 50-100% T), µm: 2.5 - 3.2; 3.4 - 6.8; 7.1 - 7.7; 8.1 - 11.1; 11..3 - 12.6
Boiling Point (typical)................. 39.5 °C.
Density (g/mL) at 25 °C (typical)........ 1.318


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR