Dichloromethane 99,8 GC, corrected for water content Stabilised, BAKER ANALYZED® HPLC HPLC, Spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Methylene chloride

Stabilization: ≥ 50 ppm stab
M.O.C. Category N

BAKR9315-03 9315-33 9315-02
CAJT9315-3CS 1912.56 CAD
CAJT9315-3 JT9315-33 JT9315-2
Dichloromethane 99,8 GC, corrected for water content Stabilised, BAKER ANALYZED® HPLC HPLC, Spectrophotometry, J.T.Baker®
Dichloromethane

Methylene Chloride HPLC For use in liquid chromatography (HPLC and UHPLC) and spectrophotometry.

Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8
Melting Pt: -95
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤233
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission, For Information Only For Information Only
Fluorescence Trace Impurities, measured as Quinine Base - at Emission For Information Only
Maximum for Impurities, For Information Only
Assay (by GC, corrected for water, exclusive of preservative) ≥99.8 %
Chloride (Cl) ≤10 ppm
Preservative (cyclohexene) ≥50 ppm
Residue after Evaporation ≤2.0 ppm
Titrable Acid (µeq/g) ≤0.3
Water (by KF, coulometric) ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR