Diethanolamine 98,5 GC, corrected for water content, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: ß,ß′-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine, 2,2'-Iminodiethanol

M.O.C. Category N

BAKR9227-03 9227-01
CAJT9227-1CS 95859.61 CAD
CAJT9227-1 CAJT9227-3 JT9227-1
Diethanolamine 98,5 GC, corrected for water content, BAKER ANALYZED® ACS
Diethanolamine

Lot analysis on label.

Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
Boiling Pt: 268.8
Melting Pt: 28
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 176
Storage Temperature: Ambient
MDL Number: MFCD00002843
CAS Number: 111-42-2
EINECS: 203-868-0
REACH: 01-2119488930-28
Merck Index: 13,03134

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay ((HOCH₂CH₂)₂NH) (by GC, corrected for water) ≥98.5 %
ACS - Apparent Equivalent Weight 104.0 - 106.0
ACS - Color (APHA) ≤15
ACS - Monoethanolamine ≤1.0 %
ACS - Triethanolamine ≤1.0 %
ACS - Residue after Ignition ≤0.005 %
ACS - Heavy Metals (as Pb) ≤0.002 %
ACS - Iron (Fe) ≤0.001 %
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.15 %
Chloride (Cl) ≤5 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR