N,N-Dimethylformamide 99,8 GC, corrected for water content, PHOTREX® Spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF
M.O.C. Category N
BAKR9222-01
BAKR9222-03
CAJT9222-1CS
1119.94
CAD
CAJT9222-1
CAJT9222-3
N,N-Dimethylformamide 99,8 GC, corrected for water content, PHOTREX® Spectrophotometry, J.T.Baker®
N,N-Dimethylformamide
For spectrophotometry. UV cutoff 270nm. Lot analysis on label.
- UV cutoff 270 nm
Formula:
HCON(CH₃)₂ MW: 73.09 g/mol Boiling Pt: 153 Melting Pt: -61 Density: 0.949 g/cm³ (20 °C) Flash Pt: 57.5 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3 III REACH: 01-2119475605-32 Merck Index: 13,03269 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (HCON(CH₃)₂) (by GC, corrected for water) | ≥ 99.8 % |
Appearance | Passes Test |
Color (APHA) | ≤ 15 |
Residue after Evaporation | ≤ 0.0005 % |
Titrable Acid (meq/g) | ≤ 0.0005 |
Titrable Base (meq/g) | ≤ 0.003 |
Water (H₂O)(by Karl Fischer titrn) | ≤ 0.05 % |
Trace Impurities - Iron (Fe) | ≤ 0.1 ppm |
Optical Absorbance (1-cm path vs water) - 270 nm | ≤ 1.00 |
Optical Absorbance (1-cm path vs water) - 275 nm | ≤ 0.30 |
Optical Absorbance (1-cm path vs water) - 295 nm | ≤ 0.10 |
Optical Absorbance (1-cm path vs water) - 310 nm | ≤ 0.05 |
Optical Absorbance (1-cm path vs water) - 340 nm | ≤ 0.01 |
Optical Absorbance (1-cm path vs water) - 400 nm | ≤ 0.01 |
Product Information (not specifications): | |
Windows of Infrared Transmittance (0.1-mm path, 50-100% T), µm: | |
2.5 - 3.0; 3.8 - 5.1; 5.2 - 5.5; 10.3 - 11.4; 11.7 - 14.7 | |
Boiling Point (typical)................. 153.0 °C. | |
Density (g/mL) at 25 °C (typical)........ 0.944 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...