N,N-Dimethylformamide 99,8 GC, corrected for water content, BAKER ANANYZED™ ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF

M.O.C. Category N

BAKR9221-R BAKR9221-01 BAKR9221-33 BAKR9221-06 9221-03
CAJT9221-REA 9373.6 CAD
CAJT9221-R CAJT9221-1 CAJT9221-3 CAJT9221-33 CAJT9221-6 JT9221-3
N,N-Dimethylformamide 99,8 GC, corrected for water content, BAKER ANANYZED™ ACS
N,N-Dimethylformamide

Lot analysis on label.

Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153
Melting Pt: -61
Density: 0.9445 g/cm³ (25 °C)
Flash Pt: 57.5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3 III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (HCON(CH₃)₂) (by GC, corrected for water) ≥ 99.8 %
Appearance Passes Test
Color (APHA) ≤ 15
Residue after Evaporation ≤ 0.0050 %
Titrable Acid (meq/g) ≤ 0.0005
Titrable Base (meq/g) ≤ 0.003
Water (H₂O)(by Karl Fischer titrn) ≤ 0.1 %
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR