Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

Management of Change (MOC) category = R

BAKR1919-25 BAKR1919-07 BAKR1919-08 BAKR1919-01 BAKR1919-05 1919-09 BAKR1919-09
CAJT1919-25EA 7134.55 CAD
CAJT1919-25 CAJT1919-7 CAJT1919-8 CAJT1919-1 CAJT1919-5 JT1919-9 CAJT1919-9
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP
D(+)-Glucose

GMP manufactured.


  • GMP manufactured product


No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C: Water (H₂O) ≤1.0 %
USP - Assay (Anhydrous basis) 97.5 - 102.0 %
USP - Related substances: Maltose and isomaltose ≤0.4 %
USP - Related substances: Maltotriose ≤0.2 %
USP - Related substances: Fructose ≤0.15 %
USP - Related substances: Unspecified impurities, each ≤0.10 %
USP - Related substances: Total Impurities ≤0.5 %
USP - Color and Clarity of Solution Passes Test
USP - Dextrin Passes Test
USP - Soluble Starch, Sulfites ≤15 ppm
USP - Conductivity (uS cm-1) ≤20 µS/cm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR