Dextrose monohydrate, powder, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

BAKR1910-05 1910-01
CAJT1910-1EA 423.03 CAD
CAJT1910-1 CAJT1910-5 JT1910-1
Dextrose monohydrate, powder, BAKER ANALYZED®, J.T.Baker®
Dextrose monohydrate

Powder. Lot analysis on label.

Density: 1.54 g/cm³
CAS Number: 5996-10-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Insoluble Matter ≤0.005 %
Residue after Ignition ≤0.015 %
Chloride (Cl) ≤0.005 %
Sulfate and Sulfite (as SO₄) ≤0.005 %
Starch Passes Test
Arsenic (As) ≤0.200 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR