Dimethyl sulfoxide, GR ACS ACS, Supelco®

Supplier: MilliporeSigma

Synonyms: Methanesulphinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO, Methanesulfinylmethane

MX1458-3 MX1458-5 MX1458-6
CAMX1458-3CS 2878 CAD
CAMX1458-3 CAMX1458-5 CAMX1458-6
Dimethyl sulfoxide, GR ACS ACS, Supelco®
Dimethyl sulphoxide

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189
Melting Pt: 18.5
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Appearance Clear, colorless liquid
Color (APHA) 10 max
Residue after evaporation 0.01% max
Titrable acid 0.001meq/g max
Water (H2O) 0.1% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR