Dimethyl sulfoxide, OmniSolv®, Supelco®

Supplier: MilliporeSigma

Synonyms: Methanesulphinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO, Methanesulfinylmethane

MX1456-NP20 MX1456-1 MX1456P-1 MX1456P-6
CAMX1456-NP20EA 8494 CAD
CAMX1456-NP20 CAMX1456-1 CAMX1456P-1 CAMX1456P-6
Dimethyl sulfoxide, OmniSolv®, Supelco®
Dimethyl sulphoxide

Clear liquid. For HPLC, gas chromatography, and spectrophotometry. The infrared spectrum conforms to the standard. Filtered through a 0.2µm element. UV cutoff 263nm. Soluble in water. Certificate of Analysis on label. Flammable and toxic effects.

Formula: (CH₃)₂SO
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18.5 °C
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Color (APHA) 10 max
Residue after evaporation 5ppm max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Infrared spectrum Conforms to standard
UV Abs. at 263 nm 1.000AU max
UV Abs. at 270 nm 0.400AU max
UV Abs. at 275 nm 0.250AU max
UV Abs. at 280 nm 0.200AU max
UV Abs. at 335 nm 0.020AU max
UV Abs. at 350-400 nm 0.010AU max
Water (H2O) 0.05% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR