KIMTECH PURE® G3 White Nitrile Gloves, Kimberly-Clark
Supplier: Microflex
56883 56882 56881 56884 56880 56881 56883 56882 56886 56880
CA14214-530PK 38.34 CAD
CA14214-530 CA14215-760 CA14215-762 CA14215-764 CA14215-766 CA14215-768 14215-764 14215-760 14215-762 14215-766 14215-768
KIMTECH PURE® G3 White Nitrile Gloves, Kimberly-Clark
Gloves
Powder-free nitrile gloves have textured fingertips to facilitate control and grip.
  • Static dissipative in use
  • Ambidextrous
  • Beaded cuff
Certifications: Recommended for ISO Class 4 (FED-STD-209E Class 1/M1.5) or higher cleanroom environments. AQL 1.5.
Ordering information: Packaged in a Class 100 cleanroom. Packaged in cleanroom-compatible polyethylene, 100 gloves per double poly bag.