N,N-Dimethylformamide ≥99.8% (by GC), GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF

DX1730-20 DX1730-3 DX1730-1 DX1730-6 DX1730-3
CADX1730-20EA 9995 CAD
CADX1730-20 CADX1730-3 CADX1730-1 CADX1730-6 EM-DX1730-3
N,N-Dimethylformamide ≥99.8% (by GC), GR ACS, Supelco®
N,N-Dimethylformamide

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153 °C (lit.)
Melting Pt: −61 °C (lit.)
Density: 0.944 g/ml (lit.)
Flash Pt: 57.5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear
Assay (GC) 99.8% min
Color (APHA) 15 max
Residue after evaporation 0.005% max
Expiration date Exp 24 monhts from mfg date
Form Liquid
Free amine (as dimethylamine) 10ppm max
Titrable acid 0.0005meq/g max
Titrable base 0.003meq/g max
Water (H2O) 0.15% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR