N,N-Dimethylformamide ≥99.9% (by GC), Supelco®

Supplier: MilliporeSigma
Danger

DX1729-NP20 DX1729-1
CADX1729-36EA 12527.37 CAD
CADX1729-36 CADX1729-38 CADX1729-NP20 CADX1729-1 CADX1729-37 CADX1729-39
N,N-Dimethylformamide ≥99.9% (by GC), Supelco®
N,N-Dimethylformamide

Flammable.

Formula: HCON(CH₃)₂
Boiling Pt: 153 °C (lit.)
Melting Pt: −61 °C (lit.)
Density: 0.944 g/ml (lit.)
Flash Pt: 57.5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Expiration date Exp 24 months from mfg date
Assay (GC) 99.9% min
Appearance Clear, free from particulates
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Liquid
Free amine (as dimethylamine) 5ppm max
Residue after evaporation 5ppm max
Titrable acid 0.5µeq/g max
Titrable base 0.5µeq/g max
UV Abs. at 270 nm 1.00AU max
UV Abs. at 275 nm 0.30AU max
UV Abs. at 280 nm 0.20AU max
UV Abs. at 300 nm 0.05AU max
UV Abs. at 350 nm 0.01AU max
UV Cut-off 270nm max
Water (H2O) 300ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR